Would you like to log in again?
Egg Yolk Tellurite, 100%, Hardy Diagnostics
Supplier: Hardy Diagnostics
R450330 R450330
89407-702EA 147.8 USD InStock
89407-702 CA89407-702
Egg Yolk Tellurite, 100%, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
A sterile emulsion of egg yolk containing potassium tellurite for use in Criterion™ Baird-Parker Agar Base, 89405-106.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty