Would you like to log in again?
Malt Extract Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
W28 W80 L128 W80 W28 L128
89407-712PK 89.66 USD InStock
89407-712 CA89407-712 CA89407-718 89407-726 CA89407-726 89407-718
Malt Extract Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of fungi
For the cultivation and enumeration of yeasts and molds.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty