Would you like to log in again?
Mineral Oil
Supplier: Hardy Diagnostics
Z80 Z79 Z79
89407-734EA 25.43 USD InStock
89407-734 CA89407-734 76213-938
Mineral Oil
Media for Microbiology Dehydrated Media for Microbiology
Used as a sealant to create an anaerobic environment for biochemical tests such as decarboxylase; and oxidation and fermentation reactions.
  • For oil overlays
Can be used for filling cupules in bacterial identification strips, such as API or Microgen.
Industry: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, pharmaceutical manufacturing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty