Would you like to log in again?
Rose Bengal Agar with Chloramphenicol, Hardy Diagnostics
Supplier: Hardy Diagnostics
W87 W87
89407-862PK 102.77 USD InStock
89407-862 CA89407-862
Rose Bengal Agar with Chloramphenicol, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and enumeration of yeasts and molds
Many yeasts and molds will not grow in an acidic pH environment. This media has a neutral pH which helps grow yeast and mold, but is also conducive to the growth of bacteria. The addition of chloramphenicol helps reduce bacterial overgrowth.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty