Would you like to log in again?
Sabdex (Sabouraud Dextrose) Agar
Supplier: Hardy Diagnostics
U227 W73 Q83 H39 Q31 U175 Q75 W71 U227 W71 W74 H39 K54 G159 U353 W74 U353 Q31 G159 U73 U431 Q75 W70 U352 W70 U352 K54 Q83 U175 W73 U73 U431 L40 L40 W250
89407-898PK 116.05 USD InStock
89407-898 89407-876 89407-894 89407-872 CA89407-882 89407-874 89407-896 CA89407-884 CA89407-904 CA89407-906 CA89407-900 CA89407-868 CA89407-902 89407-180 CA89407-908 89407-880 89407-882 CA89407-874 CA89407-896 89407-900 CA89407-898 89407-868 CA89407-870 89407-884 CA89407-894 CA89407-872 89407-906 89407-908 89407-902 89407-904 CA76168-836 76169-780 89407-870 CA89407-880 CA89407-180
Sabdex (Sabouraud Dextrose) Agar
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Dextrose Agar, Sabouraud Dextrose Broth, and Sabouraud Dextrose Agar, Emmons are recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts.
Ideal for environmental monitoring in a clean room or clean bench setting.
Sabouraud Dextrose Agar with Chloramphenicol, Sabouraud Dextrose Agar with Chloramphenicol and Gentamicin, and Sabouraud Dextrose Agar with Chloramphenicol and Tetracycline are recommended for the selective isolation of fungi and yeasts from clinical and nonclinical specimens.
Caution: Not intended to be used for the diagnosis of human disease.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty