Would you like to log in again?
Selenite Cystine Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
K69 K69
89407-920PK 82.29 USD InStock
89407-920 CA89407-920
Selenite Cystine Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective enrichment of Salmonella species Cystine enhances growth of Salmonella spp.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty