Would you like to log in again?
SIM (Sulfide, Indole, Motility) Medium, Hardy Diagnostics
Supplier: Hardy Diagnostics
Q30 Q30
89407-924PK 97.78 USD InStock
89407-924 CA89407-924
SIM (Sulfide, Indole, Motility) Medium, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the identification of gram negative bacilli based on sulfide production, indole formation, and motility.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty