Would you like to log in again?
TAT Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
U318
89407-952PK 249.06 USD InStock
89407-952
TAT Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the detection of microorganisms.
For the detection of microorganisms from topical drugs, cosmetics, and other viscous materials.
Ordering information: Minimum order of 10 packs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty