Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ketogenic Diet, AIN-76A-Modified, High Fat, Paste, Bio-Serv

Supplier: Bio-Serv
S3666 S3666 F3666 F3666
89408-206EA 285.38 USD InStock
89408-206 CA89408-206
Ketogenic Diet, AIN-76A-Modified, High Fat, Paste, Bio-Serv
Animal Supplies Animal Feed
A high fat, low carbohydrate diet in a paste form.

  • Ratio of fat to carbohydrate and protein is approximately 6:1
  • Nutritionally assayed
  • Shelf Life: 6 months refrigerated

Ingredients: Lard, Butter, Corn Oil, Casein, Cellulose, Mineral Mix, Vitamin Mix, Dextrose.

Caloric value: 7.24 kcal/g
Nutritional ash content: 3%
Nutritional carbohydrate content: 3.2%
Nutritional fat content: 75.1%
Nutritional fibre content: 4.8%
Nutritional moisture content: <10%
Nutritional protein content: 8.6%

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty