Would you like to log in again?
Mouse MD's™ Baytril Tablets for Rodents, Bio-Serv
Supplier: Bio-Serv
Mouse MD's™
F05266 MD300-050 F05266 MD300-050
89408-278EA 90.98 USD InStock
CA89408-278 89408-278 CA89408-226 89408-226
Mouse MD's™ Baytril Tablets for Rodents, Bio-Serv
Animal Supplies Animal Feed
Highly palatable, nutritionally complete, grain-based, bacon-flavored tablets
Each 5 g scored tablet contains 0.5 mg of Baytril and offers accurate and flexible dosing. Mouse average dose is one 5 g tablet per 20 g of body weight.
Ingredients: Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (Preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, Calcium Silicate, Bacon Flavor, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin.
Convenient and safe alternative to traditional dosing methods. Easy to administer; place on cage floor for easy access. Tablets are formulated to meet the daily nutritional and medication requirements of rodents. More convenient and less risk of dehydration compared to using medicated water. Cold tableting process eliminates drug degradation. MD’s can be customized to meet specific requirements. Shelf Life: 6 months, cool dry storage
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty