Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

The Belly Dancer Shakers, 220 V

Supplier: IBI Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BDRAA220S BDRLS0006 BDRLS0004 BDRLS0005
10040-376EA 2867.45 USD InStock
10040-376 10040-374 89408-316 10040-372
The Belly Dancer Shakers, 220 V
Shakers and Mixers Orbital Shakers
The unique, undulating motion of The Belly Dancer shaker is designed to provide the optimal movement for tissue culture, staining and destaining gels, Northern, Southern, or Western blotting, and immunoprecipitation

  • 220 V model

The Belly Dancer has a simple, durable construction and is made of chemically resistant materials including stainless steel and anodized aluminum parts with a high bake ester finish on the case. The Belly Dancer comes with an open-sided platform to accommodate oversized containers and a removable, washable non-skid platform mat. The flat power cord can also pass through the door gasket of incubators without compromising the environment. The platform can easily be adjusted with the platform pitch control collar from level motion to an 8° pitch. The Belly Dancer can hold up to 20lbs. with extremely quiet operation. The ambient operating temperature range is 0°C to 40°C. The speed varies from 0 to 100rpm with no cogging at slow speeds.

Accessories for The Belly Dancer include a 2nd Story Platform, a Large Nesting Platform, and a Tube Mat. The 2nd Story Platform doubles the platform capacity and comes with 4.5" aluminum spacer legs and a non-skid removable mat. The Large Nesting Platform, which is 18 x 18", increases the platform capacity by 225% and comes with a non-skid removable mat. The Urethane Tube Mat is a 12 x 12" urethane mat that allows the Belly Dancer to mix the contents of small tubes up to 50ml in size.

Belly Dancer Lab Start-Up Kits are now available with your choice of accessories. The Belly Dancer with 2nd Story Platform includes The Belly Dancer and the 2nd tier platform kit. The Belly Dancer with Nesting Platform includes the Belly Dancer and the 18" x 18" nesting platform kit. The Belly Dancer with Tube Mat includes The Belly Dancer Shaker and the 12" x 12" urethane tube mat.

The simple liquid agitation system, consisting of an orbiting platform flexibly anchored at four corners so that the platform motion, when adjusted by the platform pitch control collar, is an extremely effective combination of rocking platform mixer and orbital shaker. Agitation is gentler but more efficient than the action of ordinary shakers. The smooth combination of vertical and horizontal orbital motion allows for more rapid exchange of solvents in applications like staining and destaining gels and protects sensitive cell lines from sheering in cell culture.

Caution: 220 V model. Corresponding models with voltage suitable for use in US (10040-368, 10040-370, 10040-378, 89408-314) are also available, please see separate product page.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty