Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hi/Lo Profile Rocker, 220 to 230 V

Supplier: IBI Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

ROCLS0005 ROCLS0004 ROCLS0006 ROCAA220S
89409-870PK 1956.63 USD InStock
89409-870 89408-328 89409-866 89409-868
Hi/Lo Profile Rocker, 220 to 230 V
Shakers and Mixers Rotating/Rocking Shakers
The lightweight, low-profile rocker unit is designed for versatility.

  • 220 to 230 V model

The low profile rocker fits easily into laboratory incubators, ovens, and refrigerated units without rearranging shelves or disturbing other materials. The thin, flat power cord reaches external outlets without damaging incubator or refrigerator gaskets. The platform is continuously adjustable from 2° to 11° and can be effortlessly changed in 15 seconds. The speed varies from 0 to 22 rpm with no cogging at slow speeds.

All finishes and hardware are stainless steel and the rocker can be easily dissembled for sterilizing in the autoclave. The rocker can hold a maximum load of 20 lbs. A maximum of 4 rocker platforms increases the rocker capacity using the same amount of bench space.

It operates on bench tops, in high humidity or dry incubators, in high temperature ovens (to 65 °C), and refrigerated units (to 0 °C). Typical applications include: Staining and Destaining Gels, Northern and Southern Hybridizations, Western Blotting and Rinsing Membranes, Cell Culture Incubation, and Phage Elutions or Amplification of cDNA libraries.

Certifications: The 230 V units are CE marked.

Caution: 220 to 230 V models. Corresponding models with voltage suitable for use in US (89408-326, 89409-860, 89409-862, 89409-864) are also available, please see separate product page.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty