Would you like to log in again?
Dispenser Dual Mode, PETG, TrippNT
Supplier: TrippNT
51935 51912
89422-916EA 126.46 USD InStock
89422-916 89408-484
Dispenser Dual Mode, PETG, TrippNT
Wipe Dispensers
This Clear Dual Dispensing Paper Towel Holder lets users grab C-fold paper towels from the top or bottom
Installs on the wall with tape (not included) or key holes. Made from tougher-than-acrylic PETG, it fits all sizes of multi-fold paper towels and permits taking multiple towels from the top or one single towel from the bottom. Two sizes available. Holds one stack (about 150 paper towels) or two stacks (about 300 paper towels).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty