Would you like to log in again?
p62 ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
900-212-0001
89409-178EA 760 USD InStock
89409-178
p62 ELISA Kit, Enzo Life Sciences
Assays ELISAs
Highly validated, quantitative p62 ELISA kit available for autophagy research.
For the quantitative determination of human, rat and mouse p62 in cell lysate samples. Cited sample type includes in PBMC lysates.
The p62 ELISA kit is a colorimetric, immunometric immunoassay kit with results in 3 hours.
Ordering information: Contains Microtiter Plate, Antibody, Conjugate, Assay Buffer 13, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, RIPA Cell Lysis Buffer 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty