Would you like to log in again?
LAB ARMOR BEAD BATH 20L 230V
Supplier: Sheldon Manufacturing
20 Liter Bead Bath With 15 Liter Lab Armor Beads And With Digital Microprocessor Control Compatible With Beads Only^ Overtemperature protection stainless body and lid,specify type plug.Ext. Dimensions: 12 x 18 x 6 (30.5 x 45.7 x 15.2 cm) Temperature: 5degC above ambient to 80degC.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LAB ARMOR BEAD BATH 20L 230V 89409-238 74220-720 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty