Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Lab Armor® Walkabout Trays, Shel Lab
Supplier: Lab Armor
39438-002 39438-001
89409-258EA 171.83 USD InStock
89409-258 89409-260
Lab Armor® Walkabout Trays, Shel Lab
Trays/Dishes/Pots Laboratory Trays/Dishes
The Walkabout Tray is jaw-droppingly clever
The Walkabout Tray can maintain a 5 ml liquid sample at 37°C ± 1.3°C for 30 minutes after removing it from the water bath. It is durable and lightweight, perfect for use under the hood. Molded one-piece construction, condensation free.
Does not include a lid. Hand wash with mild detergent, towel dry.
It's a quarter the size of a traditional lab bucket. Making it easy to use under the hood or in tight benchtop spaces. With its superior insulative properties, it keeps your samples and reagents at temperature after removing them from the bead bath, incubator or refrigerator.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty