Would you like to log in again?
Lab Armor® Double Bead Blocks, SHEL LAB
Supplier: Sheldon
Lab Armor®
52200-RED 52200-BLU 52200-GLD 52200-SLV 52200-BLK
89409-278EA 679.98 USD InStock
89409-278 89409-280 89409-282 89409-284 89409-286
Lab Armor® Double Bead Blocks, SHEL LAB
Block Heaters Blocks
The Bead Block™ is an aluminum block filled with 0.5L of Lab Armor Beads™ designed for use with any standard, commercially available dry bath
The Bead Block™ replaces common drilled-out aluminum heat blocks and provides optimal temperature uniformity and accuracy while incubating common lab vessels. These blocks accommodate a wide variety of vessels, including microfuge tubes, test tubes, and even microplates and slides. The Bead Block™ eliminates the need for using different size blocks to fit different sample vessels and can be easily lifted out of the dry bath unit for portable use anywhere in the lab.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty