Would you like to log in again?
SHEL LAB Refrigerated BOD Peltier Incubator, Sheldon
Supplier: Sheldon
SRI20P
89409-360EA 11808.6 USD InStock
89409-360
SHEL LAB Refrigerated BOD Peltier Incubator, Sheldon
Incubators BOD Incubators
Using innovative thermoelectric cooling technology, this model eliminates the need for a refrigeration compressor.
  • Forced convection
  • Independent over temperature thermostat and alarm
  • Adjustable leveling feet
  • One internal electrical outlet
Uses 78% less power than alternative models and reduces room air conditioning loads by 75%. The model includes 75 lb. capacity shelves, which eliminates sagging. Meets APHA specifications for Biochemical Oxygen Demand (B.O.D.) and includes a mechanical convection system to ensure even air distribution, digital temperature set controller, over temperature limit control, and a digital temperature display.
Low Temperature Incubators are ideal for testing of sewage and wastewater, as well as other low and room temperature applications.
Certifications: TUV approved. CE marked. UL listed. CSA certified.
Ordering information: Two-year limited manufacturer's warranty.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty