Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

TRACK-IT™ Rugged Temperature Logger, Monarch Instrument

Supplier: Monarch
TRACK-IT™
5396-0105 5396-0105-CAL
89409-782EA 474.95 USD InStock
89409-782 89184-090
TRACK-IT™ Rugged Temperature Logger, Monarch Instrument
Data Loggers
The rugged temperature data logger is a submersible battery powered stand alone compact data logger that records up to 32000 samples of temperature data.

  • Push button or program to start recording
  • Three year battery life
  • N.I.S.T. calibration available
  • Accurate internal clock (1 minute/year)
  • Projected time and date of full log
  • Mean kinetic temperature calculation
  • Two seconds to 24 hours sample rate

The unit is easily configured using the included Track‐It™ Software. Simply connect the logger to an open USB port on your PC and the software automatically identifies the logger. Use the internal push button to start and stop recording and check status or configure the unit to start recording immediately, start and/or stop at a predetermined time and date, or only when an alarm condition is sensed.

The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery. High accuracy and repeatability. Wide temperature range −40 to 257 °F/125 °C.

Stainless steel IP68 packaging make it versatile and rugged enough for the more strenuous temperature logging applications. Moisture, corrosion, and force are prevented from affecting your measurements, as well as shallow submersion.

LED: Indication of alarms, recording, triggered. Record on/off, reset alarms.

Dimension:3.585" (91.06L mm)×0.787" (20Ø mm)

Certifications: CE marked.

Ordering information: Interactive software, USB interface cable and three year battery included.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty