Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument
Supplier: Monarch
TRACK-IT™
5396-0304-CAL 5396-0302-CAL 5396-0303 5396-0304 5396-0303-CAL 5396-0305-CAL 5396-0301 5396-0301-CAL 5396-0305 5396-0302
89409-794EA 888.42 USD InStock
89409-794 89409-796 89409-798 89200-716 89409-800 89409-802 89184-588 89184-586 89184-584 89409-792
TRACK-IT™ Pressure and Temperature Logger, Monarch Instrument
Data Loggers
The pressure/temperature data logger is a battery powered stand alone water tight compact data logger that records up to 64000 samples of pressure and/or temperature data
The logger is packaged in a rugged submersible 316L stainless steel housing and has a standard ¹/₄" NPT fitting. The replaceable internal lithium battery has up to a 5 year life. The unit is easily configured using the included Track‐It™ software. Simply connect a standard USB cable to the logger and an open USB port on your PC and the Track‐It™ software automatically identifies the logger.
Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery.
It can be configured to record pressure and temperature or either parameter alone to maximize data storage space. The Track‐It™ pressure/temperature data logger can be ordered in four pressure ranges: 0 - 35, 0 - 150, 0 - 350 and 0 - 550 psi.
Certifications: CE marked.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty