Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
ULTRA-TURRAX® T25 Digital Disperser, IKA® Works
Supplier: IKA
3725000 0003725001
89409-882EA 3788.23 USD InStock
89409-882 89409-880
ULTRA-TURRAX® T25 Digital Disperser, IKA® Works
Homogenizers Benchtop Homogenizers
High performance dispersing instrument with digital speed display for volumes ranging from 1 to 2000mL (H2O)
Units are equipped with digital speed display, electronic speed control, error code display, and electronic overload protection. The stainless steel dispersing elements can be cleaned quickly and easily. Plastic disposable dispersing elements are available in two sizes. Quiet operation.
Reddot design award winner 2012.
It offers a wide speed range, from 3000 to 25,000rpm, that enables users to work at high circumferential speeds even with small rotor diameters. A broad choice of dispersing elements adds versatility. Applications include homogenizing waste water samples, use in laboratory reactors, dispersion tasks under vacuum/pressure, and sample preparation in medical diagnostics.
Certifications: CE certified.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Dispersing elements, boss head clamps, stands, and other accessories are sold separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty