Starter units include four round corner posts and four shelves, and add-on units include two posts, four shelves and clips.
- Starter units include four round corner posts and four shelves
- Chrome finish
- Truss-reinforced edges
Chrome finish provides improved corrosion resistance. Each shelf features truss-reinforced edges for exceptional durability and can support up to 800 lbs. when weight is evenly distributed. Posts feature groove rings in 1 increments for shelf height adjustability.
Ordering information: Shipped unassembled for compact storage.