Would you like to log in again?
R2A Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
G54 G54
89410-652PK 33.53 USD InStock
CA89410-652 89410-652
R2A Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
R2A Agar, 15x100mm plate, order by the package of 10
For the detection of heterotrophic bacteria in water samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty