Would you like to log in again?
Reactive Quencher Dyes, Biotium
Supplier: Biotium
91051 80502 80500 80501
89410-780EA 540.61 USD InStock
89410-780 89410-778 89410-776 89410-774
Reactive Quencher Dyes, Biotium
Protein Linking and Labeling Systems
Reactive quenchers that act as acceptors in fluorescence resonance energy transfer (FRET) applications.
  • Thiol- and amine-reactive quenchers
  • One Sample Fluorescence Resonance Energy Transfer (OS-FRET) and FRET quenchers
NFQ1, a non-fluorescent quencher, is used for a new type of One Sample Fluorescence Resonance Energy Transfer (OS-FRET) method employing a novel, non-fluorescent methanethiosulfonate-linked acceptor that can be reversibly coupled to a target sulfhydryl residue via a disulfide bond
The thiol-reactive quencher, CFQ520 maleimide, has a broad and intense absorption (approximately 520 nm) but no fluorescence, making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.
The amine-reactive quencher, CFQ520 succinimidyl ester, has a broad and intense absorption (approximately 520 nm) but no fluorescence, making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.
N-(6-Aminohexyl)maleimide is a cross-linking reagent that can react with an amine-reactive group and a thiol group.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty