Would you like to log in again?
Agar Powder, Bacteriological, HiMedia
Supplier: HiMedia Laboratories
GRM026P-500G GRM026P-2.5KG GRM026P-2.5KG GRM026P-500G GRM026P-5KG
89411-634EA 2296.48 USD InStock
CA89411-632 89411-634 89411-630 CA89411-630 89411-632
Agar Powder, Bacteriological, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Agar powder, Bacteriological is recommended for routine bacteriological work and particularly useful in pharmaceutical preparation where it passes in Microbial limit tests for the presence of bacteria.
Bacterial Content as per IP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty