Would you like to log in again?
Excelta Corp® Pin Vises, Brass
Supplier: Excelta
227B-ET 227B 227D-ET 227C-ET 227A-ET 227C-P 227A-ET
63043-694EA 38.27 USD InStock
63043-694 76240-088 75880-876 63041-538 89411-690 63041-542 63041-540
Excelta Corp® Pin Vises, Brass
Dissection Needles
The 277 series of pin vise provides handle for various drill bits or probes.
  • Miniature design for accessibility under scope
  • Available in two different chuck sizes
  • Serrated handle for improved gripping
  • Available with ergonomic grip
The 'Series 227' style is a brass pin vice specifically designed to handle small drills or probes. The 227A has a 0" -.015" opening chuck. The 227B has a 0 to .025" opening chuck. The 227C and 227D include a .010" replaceable probe. The suffix 'ET' features the Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty