Would you like to log in again?
Cutters, Tubing, Excelta
Supplier: Excelta
2922-14 2922-38 51-T 2922-18 2922-14-X
76250-660EA 56.38 USD InStock
76250-660 89411-842 89411-844 89411-712 75880-684
Cutters, Tubing, Excelta
Cutters
Gives a nice clean cut to a variety of soft tubing materials.
  • Medical 'clean-cut' tubing cutter
  • Replaceable blades
  • Ergonomic grips and dual leaf springs
The 2922 Series and the 51-T medical tubing cutter give a nice clean cut to a variety of soft tubing materials. The blades on the 2922 series are replaceable and the grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty