Would you like to log in again?
Tweezers, Reverse Action, Excelta Corp®
Supplier: Excelta
Four Star™
DN-2A-SA-SE 31B-SA-SE 29-S-SE 30-S-SE 31-SA-C DN-7-SA-SE DN-3C-SA-SE DN-7-SA-SE 31-SA 29-SA 30-SA 31-SA
63041-854EA 139.96 USD InStock
63041-854 10066-492 89411-748 63041-922 76238-682 63041-912 10066-488 63041-928 94024-404 89411-746 76245-188 10066-490
Tweezers, Reverse Action, Excelta Corp®
Forceps
The reverse action tweezers are closed at rest, allowing for the operator to hold an item without any finger pressure on the tweezer.
  • Will hold parts without finger pressure for extended time
  • Tool determines pressure on component, not operator
Reverse action tweezers are normally closed at rest. This allows the item to be held without hand pressure for any length of time. We offer 2 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty