Would you like to log in again?
Tweezers, Curved Tapered Relieved Tip, Style 5A, Excelta Corp®
Supplier: Excelta
5B-SA-ET 5B-SA 5A-SA-SE 5A-S 5B-SA-PI 5A-SA 5A-S 5A-SA-PI-ET 5A-S-SE 5B-SA-PI-ET 5A-SA-PI 5A-SA-ET
63042-664EA 110.55 USD InStock
63042-664 76241-172 89411-780 63042-676 63042-666 63042-678 89411-798 89411-810 63042-670 89411-872 63042-672 89411-840
Tweezers, Curved Tapered Relieved Tip, Style 5A, Excelta Corp®
Forceps
These are ideal for microscopy, featuring offset tapered tip for excellent visibility.
  • Tapered offset tips for greater visibility/accessibility
  • Very fine points for precision
  • Offered in 3 different materials
  • Three quality levels
  • Available with ergonomic grips
These angled/bent tapered ultra-fine pointed tweezer ares primarily used for microscopy. The offset angle offers easy access and is designed for good visibility. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel. The 'ET' version features our Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty