Would you like to log in again?
Tweezers, Angled Fine Style 51, Excelta Corp®
Supplier: Excelta
50-SA-PI 51-S 5C-SA 51-SA-PI-ET 51-SA-PI-ET 51-SA-PI
63042-510EA 116.7 USD InStock
63042-510 94024-284 89411-808 10066-496 63042-682 76246-178
Tweezers, Angled Fine Style 51, Excelta Corp®
Forceps
The offset oblique tips offer excellent visibility under a scope.
  • Precision points for work under a scope
  • Offset tips offer excellent visibility
  • Offset tips excellent for accessibility in tight places
The '51 Style' series 45° offset oblique angle micro-fine tipped tweezer is ideal for working under a scope. It is also available with our Ergo-Tweeze® soft cushioned 'ET' grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat. There is also a 30° offset style available. We offer 2 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty