Would you like to log in again?
Tweezers, Lens Handling®, Excelta Corp®
Supplier: Excelta
2A-SA-LH
89411-816EA 167.47 USD InStock
89411-816
Tweezers, Lens Handling®, Excelta Corp®
Forceps
Ideal for handling delicate contact lenses.
  • Designed specifically for contact lenses
  • Rubber coated tips avoid scratches
The 2A-SA-LH three star straight tapered flat pointed tweezer with rubber coated tips is ideal for handling delicate contact lens. The overall length is 4.75 and is made of anti-magnetic Neverust® stainless steel. The tips measure 0.08" wide by 0.008" high.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty