Would you like to log in again?
Tweezers, Straight Fine Tips, Style 3, Excelta Corp®
Supplier: Excelta
3-SA-ET 3-S-SE 3-S-PI 3-SA-SE 3-S 3-SA 3-SA-PI 3-SA-PI-ET 64-SA 3-SA 3-SA-ET 3-SA-PI-ET
63042-148EA 45.72 USD InStock
63042-148 89411-770 89411-824 76238-690 76238-692 63042-150 63042-152 63042-718 63042-142 63042-154 63042-144 76241-110
Tweezers, Straight Fine Tips, Style 3, Excelta Corp®
Forceps
Style 3 is ideal for precision work, including under magnification.
  • Ideal for working under magnification
  • Available in three different materials
  • Available in three different qualtiy grades
  • Ergonomic handles are standard option
  • Teflon coated tip option for non-marring
The 'Style 3' series straight very fine precision pointed tweezer is designed for work under magnification. The overall length is 4.75". These are also availabe with ergonomic handles (ET). We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty