Would you like to log in again?
Tweezers, Angulated Wire Cutting, Excelta Corp®
Supplier: Excelta
14A-GW 15A-C 52-MW 15A-GW-PB 15A-SW 15A-GW+ 55-MW 54-MW 15A-GW-S 14A-N 15A-RW+ 15A-GW-M 15A-GW-OF 15A-GW-PB 15A-GW+ 15A-RW+ 15A-RW-M 15A-RW-OF 15A-RW-ET 15A-RW-S 15A-GW-OF 15A-GW-PB 15A-RW-PB 15A-GW-S 15A-GW-ET
89411-938EA 246.49 USD InStock
89411-938 10066-450 89411-914 89411-936 63041-244 76240-454 76471-492 89411-920 89411-922 89411-882 89411-940 75880-532 89411-924 89411-926 63041-296 10066-500 10066-446 10066-448 63041-276 63042-522 76240-442 76241-106 89411-930 76240-602 63042-582
Tweezers, Angulated Wire Cutting, Excelta Corp®
Forceps
These cutting tweezers are designed for cutting of small wires or flashing under magnification.
  • Excellent for work under a scope
  • Induction hardened for longevity
  • Focal cutting point at very tip
  • Optimum flush cut
These cutting tweezers are generally designed to cut 0.010" soft wire. They are made of high grade carbon steel with flame hardened edges. The precision of the points allow for work under magnification. This tool is considered a tip cutter in that the blades meet most predominately at the tip. All of the force applied to the tweezer gets transferred to the points. We offer a variety of added features, such as medical grade (M), parallel cutting blades (PB), optimum flush cut (OF), ergonomic (ET) and stainless steel (S).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty