Would you like to log in again?
Pup Tents™ for Mice, Certified, Bio-Serv
Supplier: Bio-Serv
Pup Tents™
K3389 K3390 K3600 K3389 K3600 K3390
89412-084EA 3.14 USD InStock
CA89412-082 89412-084 CA89412-080 89412-082 CA89412-084 89412-080
Pup Tents™ for Mice, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
A shelter for pups and weanlings made of high-temp polycarbonate
Small, low profile design fits in most mouse cagings. Deflects air flow to prevent thermal stress. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty