Would you like to log in again?
Rest Stops™ Rodent Environmental Enrichment, Certified, Bio-Serv
Supplier: Bio-Serv
Rest Stops™
K3392 K3391 K3392 K3391 K3393
89412-108EA 10.06 USD InStock
89412-108 CA89412-106 89412-106 89412-104
Rest Stops™ Rodent Environmental Enrichment, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Rest Stops offer paw relief from wire bottom floors and provide a comfortable resting area
Made of transparent high-temp polycarbonate. Useful to place special needs diets or enrichment treats at cage level. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty