Would you like to log in again?
Shake-A-Treat Jr.™, Certified, Bio-Serv
Supplier: Bio-Serv
Shake-A-Treat
K3350-JR K3350-JR
89412-110EA 41.68 USD InStock
89412-110 CA89412-110
Shake-A-Treat Jr.™, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
A handheld interactive foraging device engineered to contain no adhesives
Use with a variety of small foraging treats up to 1/2" diameter. Easy-to-clean two-piece construction. Made of transparent amber high-temp polycarbonate and stainless steel. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty