Would you like to log in again?
MicroGenesis™ Wipes, Contec®
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LWMM0300 LWMM0100 LWMM0100
89412-352CS 601.13 USD InStock
CA89412-350 89412-352 89412-350
MicroGenesis™ Wipes, Contec®
Wipes Disposable Wipes and Towels Wipes for General Applications
MicroGenesis™ wipes deliver the maximum amount of microfiber to pick up and retain particles, dust, and other contaminants in cleanrooms and other critical applications.
  • Recommended for ISO Class 6-8 environments
  • Microfiber is only applied to the wiping surface creating a cost effective single use microfiber wipe
  • Microfiber fabric easily picks up and retains particles
  • Highly sorbent and suitable for use with a wide range of disinfectants
For dry applications where particle pickup and retention is critical MicroGenesis™ Wipes deliver unprecedented performance. The wipe is ideal for picking up the powder residue generated in tablet and fine powder pharmaceutical manufacturing or grease removal before manufacturing batches.
This particle pickup makes the wipe useful for surface preparation prior to painting, laminating, or other finishing applications. In addition, unique processing gives the MicroGenesis™ Wipe excellent sorbency characteristics, making it ideal for multiple applications and industries. It is safe for use with all disinfectants including chlorine and quaternary ammonium-based products.
MicroGenesis™ wipes are not recommended for applications with rough or perforated surfaces.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty