Would you like to log in again?
AlphaTec® 58-335, Nitrile Gloves, with AQUADRI® Technology, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
114020 114022 114019 284975 284978 114021 284979 114023 284977 284976
89413-070CS 1369.59 USD InStock
89413-070 CA89413-070 89413-072 CA89413-072 89413-064 89413-066 CA89413-064 89413-068 CA89413-066 CA89413-068
AlphaTec® 58-335, Nitrile Gloves, with AQUADRI® Technology, Ansell
Gloves
Ansell’s new, proprietary AQUADRI® technology delivers the properties of a sponge with great moisture absorption capacity, featuring AquaDri® Nitrile Foam Lining, thus keeping hands drier for a longer period of time resulting in improved comfort and performance.
  • Superior puncture and abrasion protection
  • Improved dry and wet grip
  • Gauntlet cuffs prevent dripping onto forearms
  • Great moisture absorption capacity
These gloves are designed to deliver optimal results in wet or dry work environments where chemical protection is crucial.
Ideal uses are changing oil, fixing pumps, chemical mixing/handling, cleaning, opening/draining pumps and valves, painting, applying sealants and sampling/testing.
The high-performance nitrile compound offers superior snag, puncture and abrasion protection over rubber or neoprene gloves. The diamond palm grip provides improved dry and wet grip while the foldable gauntlet cuff gutter prevents dripping on the forearm.
Ordering information: Available in a shorter 12" length, see AlphaTec 58-330.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty