Would you like to log in again?
ART® Barrier Reload System, Thermo Scientific
Supplier: Thermo Scientific ART
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ART®
2149P-RT 2139-RT 2070-05-RT 2139-05-RT 2179-05-RT 2070-RT 2069-RT 2069-05-RT 2179-RT 2149P-05-RT
89413-270CS 874.66 USD InStock
89413-270 89413-292 89413-184 89413-294 89413-186 89413-232 89413-266 89413-212 89413-234 89413-214
ART® Barrier Reload System, Thermo Scientific
Pipette Tips
ART® portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in the laboratory.
Due to the established level of protection, researchers have made ART® self-sealing barrier tips their first choice in sample protection. ART® barrier tips provide 100% security against aerosols and liquid contamination. The space saving, stackable tip packaging allows for tips to pass through offset holes in lower insert. The easy to use blister sleeve guide rails align tip inserts for a smooth transfer into the rack. Supplied in sterile packaging, the outer sleeve protects tips from exterior contamination while refilling inserts. The environmentally-friendly tip packaging option reduces plastic waste. The rigid insert design provides reliable tip loading into multichannel devices. All tips are traceable, with lot number and description printed on every insert.
Renowned for reliable quality, the ART® series of pipette tips is now offering a complete selection of Barrier, Non-Filter, and Specialty tips. All formats deliver proven quality, performance, and reliability, backed by the highest Thermo Scientific standards and worldwide support.
Certifications: RNase-, DNase-, DNA-, ATP-, and pyrogen-free.
Ordering information: An empty rack is required for Reload Tower use.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty