Would you like to log in again?
Letheen Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
U39
89413-494PK 121.02 USD InStock
89413-494
Letheen Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Letheen Broth to determine bactericidal activity of quaternary ammonium compounds.
For use to determine bactericidal activity of quaternary ammonium compounds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty