Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barnstead™ GenPure™ xCAD Plus Water Purification Systems, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

50136172 50136152 50136172 50136170 50136151 50136170 50136169 50136153 50136171 50136171 50136146 50136151 50136153 50136152 50136169 50136146
89414-728EA 14560 USD InStock
CA89414-728 CA89414-716 89414-728 CA89414-736 89414-716 CA89414-724 CA89414-734 89414-724 89414-736 CA89414-722 CA89414-732 89414-722 CA89414-720 89414-734 89414-720 89414-732
Barnstead™ GenPure™ xCAD Plus Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead GenPure xCAD Plus water purification systems provides up to 200L/day of Type 1, ultrapure (18.2 megohm) water from pre-treated feed water

The GenPure xCAD Plus systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. The xCAD remote dispenser can be mounted on the wall or on the bench for maximum flexibility. In addition, up to two additional remote dispensers can be added to create up to three work stations from one water system.

The GenPure xCAD systems are offered in a variety of configurations to meet a wide range of applications needs.
The GenPure xCAD systems are ideal for atomic absorption, ion chromatography or other analytical procedures.
The GenPure xCAD UV systems are ideal for inorganic and organic trace analysis, HPLC, or ICP-MS.
The GenPure xCAD UF systems are ideal for microbiology, IVF, or monoclonal antibody applications.
The GenPure xCAD UV/UF systems are ideal for molecular biology, PCR, DNA work, cell or tissue culture media.
In addition the UV and UV/UF models can be ordered with an optional TOC monitor for applications that require TOC monitoring in addition to TOC removal. Both models with TOC monitoring include UV intensity monitoring to ensure TOC readings are accurate.

All GenPure xCAD systems include: feed water monitoring to protect your consumables, quick discconnects to make changing consumables easy, all core consumables and tubing, RS232 port, full control xCAD remote dispenser, and volumetric dispensing capability.

Remote dispenser working radius: 80 cm (32")

Certifications: CSA, CE.

Ordering information: Units come standard with universal power supply (90-240V, 50.60Hz), all necessary tubing to connect to water inlet supply. Feed water must be pre-treated source such as deionized, distilled or treated by reverse osmosis. Alternative units are available that can make Type 1 water from tap water if a source of pre-treated feedwater is not available.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES


Services: Thermo Scientific provides several service options for Thermo Barnstead Water System such as Installation, Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Barnstead Water System Technical Services" in the VWR search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST™@vwr.com for further details.

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty