Would you like to log in again?
Barnstead™ GenPure™ Pro Water Purification Systems, Thermo Scientific
Supplier: Thermo Scientific
50131952 50131922 50131922 50131948 50131950
89414-740EA 15560 USD InStock
89414-740 CA89414-748 89414-748 89414-746 89414-744
Barnstead™ GenPure™ Pro Water Purification Systems, Thermo Scientific
Water Purification Systems Pure and Ultra Pure Water Systems
The Barnstead GenPure Pro water purification systems provide up to 200L/day of Type 1, ultrapure (18.2 megohm) water from pre-treated feed water
The GenPure Pro systems offer highly versatile mounting options. The units can be mounted on the wall, on the bench or under the bench. In addition the GenPure Pro offer highly flexible dispensing with a its flexible dispensing arm to accommodate the filling of a wide range of vessels.
The GenPure Pro systems are offered in a variety of configurations to meet a wide range of applications needs. The GenPure Pro systems are ideal for atomic absorption, ion chromatography or other analytical procedures. The GenPure Pro UV systems are ideal for inorganic and organic trace analysis, HPLC, or ICP-MS.
The GenPure Pro UF systems are ideal for microbiology, IVF, or monoclonal antibody applications. The GenPure Pro UV/UF systems are ideal for molecular biology, PCR, DNA work, cell or tissue culture media. In addition the UV and UV/UF models can be ordered with an optional TOC monitor for applications that require TOC monitoring in addition to TOC removal. Both models with TOC monitoring include UV intensity monitoring to ensure TOC readings are accurate.
All GenPure Pro systems include feed water monitoring to protect your consumables, built in leak detection, quick disconnects to make changing consumables easy, all core consumables and tubing, and RS232 port.
Certifications: CSA, CE.
Ordering information: Units come standard with universal power supply (90-240V, 50.60Hz), all necessary tubing to connect to water inlet supply. Feed water must be pre-treated source such as deionized, distilled or treated by reverse osmosis. Alternative units are available that can make Type 1 water from tap water if a source of pre-treated feedwater is not available.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: Thermo Scientific provides several service options for Thermo Barnstead Water System such as Installation, Extended Warranty, Extended Warranty plus on-site Annual Preventative Maintenance, and Total Care. To learn more about these services please search "Thermo Barnstead Water System Technical Services" in the website search field.
In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST@vwr.com for further details.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty