Would you like to log in again?
COVER BEARD/ BOUFFANT CAP UNISIZE PP WHT
Supplier: CT INTERNATIONAL
Spunbound Polypropylene Bouffant Caps And Beard Covers^ Uni-Size. Color: white. Length: 38.1cm. Width: 22.9cm. Strong and durable. Controls contamination. Lightweight material offers comfort and breathability, while maintaing snug full coverage. Individually inspected. Lot traceability.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COVER BEARD/ BOUFFANT CAP UNISIZE PP WHT 89414-932 HBC1000 Case of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty