Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FAM59A Rabbit Polyclonal Antibody

Supplier: ProSci
5479
89417-224EA 531.09 USD InStock
89417-224
Anti-FAM59A Rabbit Polyclonal Antibody
Antibodies
FAM59A Antibody: FAM59A, also known as GAREM, was initially identified through a mass spectroscoptic method that measures different phosphotyrosine states of proteins in response to different cell stimuli. FAM59A is a downstream molecule in the EGF signaling pathway that is tyrosine-phosphorylated, and this phosphorylation is needed for the binding of Grb2, an adaptor protein crucial to the transduction of growth signals from the plasma membrane to the nucleus. ERK activation in response to EGF stimulation is regulated by FAM59A in COS-7 and HeLa cells, and the overexpression of FAM59A stimulated cell proliferation and colony formation in soft agar, suggesting that FAM59A might be a critical protein with roles in ligand-mediated signaling pathway of the EGF receptor and the tumorigenesis of cells.

Type: Primary
Antigen: FAM59A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty