Would you like to log in again?
Sani-Cloth® AF3 Germicidal Disposable Wipes, PDI®
Supplier: PDI
PDIP13872
89422-660CS 396.51 USD InStock
89422-660
Sani-Cloth® AF3 Germicidal Disposable Wipes, PDI®
Wipes Disposable Wipes and Towels Disinfectant Wipes
Sani-Cloth ®AF3 surface disinfectant cleaner germicidal wipes alcohol and fragrance free formula ideal for use around patients, residents and staff, especially those with respiratory sensitivities.
  • Effective against 45 microorganisms in 3 min
  • Bactericidal, tuberculocidal, virucidal
  • Multi-count canister accommodates frequent usage
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty