Would you like to log in again?
Corning® Trace Elements B, Corning
Supplier: Corning
Corning®
25-022-CI 25-022-CI
89422-908EA 72.26 USD InStock
89422-908 CA89422-908
Corning® Trace Elements B, Corning
Media for Microbiology Additives and Supplements for Microbiology
Trace Elements B Solution has the same composition as Cleveland's Trace Element I.
The combination of Trace Elements B contains the trace elements of MCDB 104 and of MCDB 301.
This product is proprietary and not available from other major suppliers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty