Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LabLink Surface Mount Raceway, Versa-Duct, PG LifeLink

Supplier: PG LifeLink
VRW130BC-108 VRW130BC-84 VRW130BC-72 VRW130BC-120 VRW130BC-96 VRW252BC-48 VRW252BC-120 VRW130BC-48 VRW252BC-72 VRW252BC-108 VRW130BC-60 VRW252BC-84 VRW252BC-96 VRW252BC-60
89422-928EA 355.02 USD InStock
89422-928 89422-938 89422-948 89422-926 89422-936 89422-924 89422-946 89422-934 89422-944 89422-932 89422-942 89422-930 89422-940 89422-950
LabLink Surface Mount Raceway, Versa-Duct, PG LifeLink
Electrical Sockets
LabLink Surface Mount Raceway (Versa-Duct), made from anodized extruded aluminum, offers a superior design to combine electrical and/or data into one unified system.

  • Extruded from #6063-T5 Aluminum Alloy
  • Single Channel Assembly (1³/₄×2³/₄×48 – 120")
  • Double Channel Assembly (2¹/₄×5¹/₄×48 – 120")
  • Single Circuit Configuration
  • 12" Pigtails are provided for Electrical Connections
  • Clear Satin Anodized Finish is Durable and Corrosion Resistant
  • 12" Removable Cover Plates for Easy Access in the Field
  • 20 Amp (NEMA-5-20R) Grey Duplex Receptacles
  • Grey Communication Holder with 2 Cat 5e Data Jacks (not wired)
  • C-UL Listed

Factory pre-cut and pre-wired to standard lengths. Single channel raceway is configured to include power receptacles and dual channel raceway will include power and data. The raceway is provided in a clear satin anodized finish. Versa-Duct multi-outlet assemblies save you time and money by reducing on-site labor costs by up to 70%. Whether designing a raceway system for your laboratory work bench or a surface mounted system for an entire building, Versa-Duct is the raceway for you. Cover with two end plates.

Certifications: cUL Listed.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty