Would you like to log in again?
Kimtech™ G3 EvT Nitrile Gloves, Kimberly-Clark Professional
Supplier: Kimberly-Clark
Kimtech™ G3
38702 38703 38700 38704 38701 38701 38703 38702 38705 38705 38704 38700
89423-030CS 1960.32 USD InStock
CA89423-040 89423-030 89423-032 CA89423-030 89423-040 89423-038 CA89423-036 CA89423-038 CA89423-032 89423-034 CA89423-034 89423-036
Kimtech™ G3 EvT Nitrile Gloves, Kimberly-Clark Professional
Gloves
In the cleanroom environment, one small particle can mean big trouble. One spec of dust on a worker’s glove can cause costly damage, therefore it's critical to have a glove with low particles and extractables, such as the Kimtech™ G3 EvT nitrile gloves.
  • Beaded cuffs for easy donning
  • Low particles and extractables
  • Not made with silicone or natural rubber latex
Kimtech™ G3 EvT nitrile gloves are designed with 'Extra-Value Technology' to have low particles and extractables. They are cleaned to meet the needs of a critical cleanroom environment. Durable and powder-free.
Gloves have excellent overall tactile performance. They are not made with silicone or natural rubber latex. Additionally, the textured and tacky surface provides increased dexterity for critical tasks in semiconductor, electronics, and medical device manufacturing facilities. They have an AQL of 1.5 for pinholes and can even be recycled.
Certifications: Exceeds the ASTM D 5151 standard for critical defects or pinholes.Recommended for ISO Class 3 and higher environments.Meets U.S. FDA regulations for food-handling applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty