Would you like to log in again?
Listeria Selective Supplement, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
SR140E
89423-156EA 582.09 USD InStock
89423-156
Listeria Selective Supplement, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
A selective supplement for the isolation of Listeria.
A selective supplement for the isolation of Listeria when used with Listeria Selective Agar Base (Oxford formulation) CM856B. Vial contents (each vial is sufficient for 500ml of medium): Cycloheximide 200mg Colistin sulphate 10mg Acriflavine 2.5mg Cefotetan 1.0mg Fosfomycin 5.0mg
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty