Would you like to log in again?
Oxoid Campylobacter Supplement, Hardy Diagnostics
Supplier: Hardy Diagnostics
SR69E SR69E
89423-160EA 315.15 USD InStock
CA89423-160 89423-160
Oxoid Campylobacter Supplement, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
A selective supplement for the isolation of campylobacters.
A selective supplement for the isolation of campylobacters when used with Blood Agar Base No.2, Columbia Agar, or Brucella Agar Bases. Vial contents (each vial is sufficient for 500ml of medium): Vancomycin 5mg Trimethoprim 2.5mg Polymyxin B 1250 IU
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty